Skip to Content
Merck
All Photos(3)

Key Documents

A75684

Sigma-Aldrich

2-Amino-5-methylpyridine

99%

Synonym(s):

2-Amino-5-picoline, 6-Amino-3-picoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
107050
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

crystals

bp

227 °C (lit.)

mp

76-77 °C (lit.)

SMILES string

Cc1ccc(N)nc1

InChI

1S/C6H8N2/c1-5-2-3-6(7)8-4-5/h2-4H,1H3,(H2,7,8)

InChI key

CMBSSVKZOPZBKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tatiana Borodina et al.
The Journal of pharmacy and pharmacology, 70(9), 1164-1173 (2018-06-30)
Anxiolytic drug zolpidem was incorporated into the microcontainers based on mesoporous calcium carbonate particles modified by diethylaminoethyl-dextran/hyaluronic acid shell. The release of zolpidem in saline solution and in polymer film modelling nasal mucosa was investigated. The anxiolytic effect of zolpidem
Hanna Rostkowska et al.
The journal of physical chemistry. A, 121(37), 6932-6941 (2017-08-30)
The UV-induced thione → thiol phototautomeric reaction has been studied for monomeric 3-thio-1,2,4-triazole (3-ST) isolated in low-temperature Ar and n-H
Jonita Stankevičiūtė et al.
Scientific reports, 6, 39129-39129 (2016-12-17)
Pyridinols and pyridinamines are important intermediates with many applications in chemical industry. The pyridine derivatives are in great demand as synthons for pharmaceutical products. Moreover, pyridines are used either as biologically active substances or as building blocks for polymers with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service