933465
ESD Epoxy Coated Tweezers
Style 2A, Stainless Steel
Synonym(s):
grippers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41111761
NACRES:
NB.35
Recommended Products
Related Categories
General description
EMS techware
Application
Epoxy coated tweezers are used in applications which require to hold a component firmly.
Electrostatic Discharge (ESD) epoxy coated tweezers with flat, accurate, round tips have the following specifications:
Length: 125mm With flat, accurate, round tips. Overall Length (OAL): 125 mm
Electrostatic Discharge (ESD) epoxy coated tweezers with flat, accurate, round tips have the following specifications:
Length: 125mm With flat, accurate, round tips. Overall Length (OAL): 125 mm
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service