900077
Vitamin K2 (MK-9)-4′,5,6,7,8,8′-13C6
≥99 atom % 13C, ≥95% (CP)
Synonym(s):
Menaquinone-9-4′,5,6,7,8,8′-13C6
About This Item
Recommended Products
isotopic purity
≥99 atom % 13C
Quality Level
Assay
≥95% (CP)
form
powder
technique(s)
mass spectrometry (MS): suitable
mass shift
M+6
storage temp.
−20°C
SMILES string
O=C1C(C)=C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CCC=C(C)C)C([13C]2=[13C][13C]=[13C][13C]=[13C]21)=O
InChI
1S/C56H76O2/c1-42(2)22-14-23-43(3)24-15-25-44(4)26-16-27-45(5)28-17-29-46(6)30-18-31-47(7)32-19-33-48(8)34-20-35-49(9)36-21-37-50(10)40-41-52-51(11)55(57)53-38-12-13-39-54(53)56(52)58/h22,24,26,28,30,32,34,36,40H,14-21,23,25,27,29,31,33,35,37,41H2,1-11H3/b43-24+,44-26+,45-28+,46-30+,47-32+,48-34+,49-36+,50-40+/i12+1,13+1,38+1,39+1,53+1,54+1
InChI key
HEKVBSATHCAGSV-PESLFLFUSA-N
Related Categories
General description
Vitamin K2 (MK-9)-4′,5,6,7,8,8′-13C6 is a vitamin K2 isotope wherein C-4′, C-5, C-6, C-7, C-8, and C-8′ carbons are replaced by 13C6.
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service