808369
Fenitrothion-(dimethoxy-13C2)
99 atom % 13C, 97% (CP)
About This Item
Recommended Products
isotopic purity
99 atom % 13C
Assay
97% (CP)
form
liquid
technique(s)
mass spectrometry (MS): suitable
mass shift
M+2
storage temp.
2-8°C
SMILES string
S=P(O[13CH3])(O[13CH3])C1=CC(C)=C([N+]([O-])=O)C=C1
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service