778885
(11R,12R)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine
≥95.0% (HPLC)
Synonym(s):
(R,R)-11,12-Diamino-9,10-dihydro-9,10-ethanoanthracene
About This Item
Recommended Products
Assay
≥95.0% (HPLC)
95.0-105.0% (NT)
form
powder
optical purity
enantiomeric excess: ≥97.0%
impurities
≤5% water
SMILES string
N[C@@H]1[C@@H](N)[C@@H]2c3ccccc3[C@H]1c4ccccc24
InChI
1S/C16H16N2/c17-15-13-9-5-1-2-6-10(9)14(16(15)18)12-8-4-3-7-11(12)13/h1-8,13-16H,17-18H2/t13-,14+,15-,16-/m0/s1
InChI key
NWDYSRZJOLDMRE-FZKCQIBNSA-N
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service