Recommended Products
isotopic purity
98 atom % D
form
solid
mp
119-122 °C (dec) (lit.)
mass shift
M+12
SMILES string
[2H][C@]1(O[2H])[C@]([2H])(O[2H])[C@](C([2H])([2H])O[2H])(O[2H])OC([2H])([2H])[C@]1(O[2H])[2H]
InChI
1S/C6H12O6/c7-2-6(11)5(10)4(9)3(8)1-12-6/h3-5,7-11H,1-2H2/t3-,4?,5+,6-/m0/s1/i1D2,2D2,3D,4D,5D,7D,8D,9D,10D,11D
InChI key
LKDRXBCSQODPBY-VVGOKITGSA-N
Related Categories
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service