Skip to Content
Merck
All Photos(1)

Key Documents

719455

Sigma-Aldrich

2-Bromomethylphenylboronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13BBrNO4
CAS Number:
Molecular Weight:
325.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

mp

196-202 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2ccccc2CBr

InChI

1S/C12H13BBrNO4/c1-15-7-11(16)18-13(19-12(17)8-15)10-5-3-2-4-9(10)6-14/h2-5H,6-8H2,1H3

InChI key

GUOSLMTXTQNEPM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service