Skip to Content
Merck
All Photos(1)

Key Documents

717533

Sigma-Aldrich

Chloro(1,5-cyclooctadiene)methylpalladium(II)

97%

Synonym(s):

(COD)PdMeCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15ClPd
CAS Number:
Molecular Weight:
265.09
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

124-129 °C

SMILES string

C[Pd]Cl.C1CC=CCCC=C1

InChI

1S/C8H12.CH3.ClH.Pd/c1-2-4-6-8-7-5-3-1;;;/h1-2,7-8H,3-6H2;1H3;1H;/q;;;+1/p-1/b2-1-,8-7-;;;

InChI key

PPNGAOYQLFBHIK-PHFPKPIQSA-M

Application

Catalyst for:
  • Addition polymerization reactions
  • Double carbonylation
  • Cyclization
  • Cyclometalation

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service