Skip to Content
Merck
All Photos(1)

Key Documents

707198

Sigma-Aldrich

Sodium dichloroacetate-13C2

99 atom % 13C

Synonym(s):

Dichloroacetic acid-13C2 sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl213CH13CO2Na
CAS Number:
Molecular Weight:
152.91
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

powder

mp

198 °C (dec.)

mass shift

M+2

SMILES string

[Na+].[O-][13C](=O)[13CH](Cl)Cl

InChI

1S/C2H2Cl2O2.Na/c3-1(4)2(5)6;/h1H,(H,5,6);/q;+1/p-1/i1+1,2+1;

InChI key

LUPNKHXLFSSUGS-AWQJXPNKSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service