Skip to Content
Merck
All Photos(1)

Key Documents

675768

Sigma-Aldrich

Oleoyl-13C18 coenzyme A lithium salt

99 atom % 13C, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C18C21H68N7O17P3S · xLi+
Molecular Weight:
1049.85 (free acid basis)
UNSPSC Code:
12352005
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

Assay

95% (CP)

form

solid

mass shift

M+18

storage temp.

−20°C

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Other Notes

Molecular weight and empirical formula based on the free acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service