609218
Fmoc-Trp-OH-α-15N
98 atom % 15N
Synonym(s):
α-N-Fmoc-L-tryptophan-α-15N1, N-(9-Fluorenylmethoxycarbonyl)-L-tryptophan-α-15N
About This Item
Recommended Products
isotopic purity
98 atom % 15N
mp
182-185 °C (lit.)
application(s)
peptide synthesis
functional group
Fmoc
mass shift
M+1
SMILES string
OC(=O)[C@H](Cc1c[nH]c2ccccc12)[15NH]C(=O)OCC3c4ccccc4-c5ccccc35
InChI
1S/C26H22N2O4/c29-25(30)24(13-16-14-27-23-12-6-5-7-17(16)23)28-26(31)32-15-22-20-10-3-1-8-18(20)19-9-2-4-11-21(19)22/h1-12,14,22,24,27H,13,15H2,(H,28,31)(H,29,30)/t24-/m0/s1/i28+1
InChI key
MGHMWKZOLAAOTD-NCGCNWTHSA-N
Related Categories
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service