603538
1-Phenylethanol-2-13C
99 atom % 13C, 99% (CP)
Synonym(s):
sec-Phenethyl-β−13C alcohol
About This Item
Recommended Products
isotopic purity
99 atom % 13C
Assay
99% (CP)
bp
204 °C/745 mmHg (lit.)
mp
19-20 °C (lit.)
density
1.020 g/mL at 25 °C
mass shift
M+1
SMILES string
[13CH3]C(O)c1ccccc1
InChI
1S/C8H10O/c1-7(9)8-5-3-2-4-6-8/h2-7,9H,1H3/i1+1
InChI key
WAPNOHKVXSQRPX-OUBTZVSYSA-N
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
187.9 °F - Pensky-Martens closed cup
Flash Point(C)
86.6 °C - Pensky-Martens closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service