Skip to Content
Merck
All Photos(1)

Key Documents

601500

Sigma-Aldrich

Lithium-7Li2 sulfate

99 atom % 7Li

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
7Li2SO4
CAS Number:
Molecular Weight:
110.09
UNSPSC Code:
12352302
PubChem Substance ID:

isotopic purity

99 atom % 7Li

form

solid

mass shift

depleted

SMILES string

[7Li+].[7Li+].[O-]S([O-])(=O)=O

InChI

1S/2Li.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2/i2*1+0;

InChI key

INHCSSUBVCNVSK-ONPSQTMSSA-L

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mareike Hütten et al.
PloS one, 9(8), e104564-e104564 (2014-08-12)
Fibrous tissue growth and loss of residual hearing after cochlear implantation can be reduced by application of the glucocorticoid dexamethasone-21-phosphate-disodium-salt (DEX). To date, sustained delivery of this agent to the cochlea using a number of pharmaceutical technologies has not been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service