Skip to Content
Merck
All Photos(2)

Key Documents

447528

Sigma-Aldrich

11-Mercapto-1-undecanol

97%

Synonym(s):

11-Hydroxy-1-undecanethiol, 11-Hydroxy-1-undecanthiol, 11-Hydroxyundecane-1-thiol, 11-Mercaptoundecanol, MUD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)11OH
CAS Number:
Molecular Weight:
204.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

solid

mp

33-37 °C (lit.)

SMILES string

OCCCCCCCCCCCS

InChI

1S/C11H24OS/c12-10-8-6-4-2-1-3-5-7-9-11-13/h12-13H,1-11H2

InChI key

ULGGZAVAARQJCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in a two-step synthesis of unsymmetrical disulfides which does not require alcohol protection.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 363-363 (2007)
Sergii Kravchenko et al.
The Analyst, 145(2), 656-666 (2019-12-05)
Complex loadings that appear on a surface with flexible spatial organisation can reveal anti-Sauerbrey behaviour due to their variable interfacial architecture even for an ultrathin monomolecular sensitive layer. The presented results demonstrate that compounds with branched and flexible structures may
Xiaochuan Huang et al.
Environmental science & technology, 54(23), 15395-15404 (2020-10-17)
Scale formation is an important challenge in water and wastewater treatment systems. However, due to the complex nature of membrane surfaces, the effects of specific membrane surface characteristics on scale formation are poorly understood. In this study, the independent effect
Tesfalem Geremariam Welearegay et al.
Nanotechnology, 29(6), 065603-065603 (2017-12-06)
Here, we report for the first time the fabrication of ligand-functionalised ultrapure monodispersed metal nanoparticles (Au, Cu, and Pt) from their pure metal precursors using the advanced gas deposition technique. The experimental conditions during nanoparticle formation were adjusted in order
M Alles et al.
Biofouling, 31(5), 469-480 (2015-07-15)
Fouling release (FR) coatings are increasingly applied as an environmentally benign alternative for controlling marine biofouling. As the technology relies on removing fouling by water currents created by the motion of ships, weakening of adhesion of adherent organisms is the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service