Skip to Content
Merck
All Photos(2)

Key Documents

376760

Sigma-Aldrich

(1,1-Dimethoxyethyl)benzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(OCH3)2CH3
CAS Number:
Molecular Weight:
166.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.49 (lit.)

bp

95-98 °C/12 mmHg (lit.)

density

1.008 g/mL at 25 °C (lit.)

SMILES string

COC(C)(OC)c1ccccc1

InChI

1S/C10H14O2/c1-10(11-2,12-3)9-7-5-4-6-8-9/h4-8H,1-3H3

InChI key

XKSUVRWJZCEYQQ-UHFFFAOYSA-N

General description

(1,1-Dimethoxyethyl)benzene is reported to participate in the bismuth triflate catalyzed deprotection of acetals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc D Carrigan et al.
The Journal of organic chemistry, 67(3), 1027-1030 (2002-02-22)
Bismuth triflate is a highly efficient catalyst (0.1-1 mol %) for the deprotection of acetals and ketals. The procedure is very facile and selective for acetals derived from ketones and conjugated aldehydes. tert-Butyldimethylsilyl ethers are stable to the reaction conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service