All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.4213 (lit.)
bp
112-113 °C (lit.)
density
0.887 g/mL at 25 °C (lit.)
SMILES string
CC1=NC(C)(C)CO1
InChI
1S/C6H11NO/c1-5-7-6(2,3)4-8-5/h4H2,1-3H3
InChI key
HZRZMHNRCSIQFT-UHFFFAOYSA-N
Related Categories
Application
2,4,4-Trimethyl-2-oxazoline was used in the synthesis of 2,2-dimethyloxazolines of C2-elongated fatty acids.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
55.4 °F - closed cup
Flash Point(C)
13 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dmitry V Kuklev et al.
Chemistry and physics of lipids, 144(2), 172-177 (2006-10-20)
Three fatty acids were synthesized from commercially available alpha-linolenic, stearidonic and eicosapentaenoic acids by C2-elongation using a four step preparative technique. The parent fatty acid methyl esters were reduced to alcohols with LiAlH(4), converted to bromides by treatment with triphenylphosphine
Po-Jung Tsai et al.
Inflammation, 41(4), 1200-1214 (2018-03-29)
Juniperonic acid (JPA; Δ5,11,14,17-20:4), originally identified in certain gymnosperm seeds, is a rare n-3 polyunsaturated fatty acid (PUFA) with lipid-modulating effects on rats and anti-proliferative effects on fibroblast cell proliferation. However, little is known how JPA exerted its immunosuppressive effect.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service