Merck
  • Ethanol promotes thiamine deficiency-induced neuronal death: involvement of double-stranded RNA-activated protein kinase.
Ethanol promotes thiamine deficiency-induced neuronal death: involvement of double-stranded RNA-activated protein kinase.
Alcoholism, clinical and experimental research (2009-04-23)
Zun-Ji Ke, Xin Wang, Zhiqin Fan, Jia Luo
ABSTRACT
Heavy alcohol consumption causes cerebellar degeneration, and the underlying mechanism is unclear. Chronic alcoholism is usually associated with thiamine deficiency (TD) which is known to induce selective neurodegeneration in the brain. However, the role of TD in alcohol-induced cerebellar degeneration remains to be elucidated. The double-stranded RNA-activated protein kinase (PKR) is a potent antiviral protein. Viral infection or binding to dsRNA causes PKR autophosphorylation and subsequent phosphorylation of the alpha-subunit of eukaryotic translation factor-2alpha, leading to inhibition of translation or apoptosis. PKR can also be activated by cellular stresses. In this study, we used an in vitro model, cultured cerebellar granule neurons (CGNs), to investigate the interaction between TD and ethanol and evaluate the contribution of their interaction to neuronal loss. TD was induced by treatment with amprolium in association with ethanol. Cell viability was determined by 3-(4,5-dimethylthiazol-2yl)-2,5-diphenyl tetrazolium bromide assay. PKR expression/phosphorylation and subcellular distribution was analyzed with immunoblotting and immunocytochemistry. Thiamine deficiency caused death of CGNs but ethanol did not. However, TD plus ethanol induced a much greater cell loss than TD alone. TD-induced PKR phosphorylation and ethanol exposure significantly promoted TD-induced PKR phosphorylation as well as its nuclear translocation. A selective PKR inhibitor not only protected CGNs against TD toxicity, but also abolished ethanol potentiation of TD-induced loss of CGNs. Ethanol promoted TD-induced PKR activation and neuronal death. PKR may be a convergent protein that mediates the interaction between TD and ethanol.
Sigma-Aldrich
PKR Inhibitor, The PKR Inhibitor, also referenced under CAS 608512-97-6, controls the biological activity of PKR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Amprolium hydrochloride
Supelco
Amprolium hydrochloride, VETRANAL®, analytical standard