Merck
  • Antiseptic activity and phenolic constituents of the aerial parts of Vitex negundo var. cannabifolia.
Antiseptic activity and phenolic constituents of the aerial parts of Vitex negundo var. cannabifolia.
Molecules (Basel, Switzerland) (2010-11-23)
Tie-Jun Ling, Wei-Wei Ling, Yuan-Jun Chen, Xiao-Chun Wan, Tao Xia, Xian-Feng Du, Zheng-Zhu Zhang
ABSTRACT
Four phenolics, salviaplebeiaside (1), γ-tocopherol (2), chrysosplenol-D (4), and isovitexin (5), along with α-tocoquinone (3) and β-sitosterol (6) were isolated from the aerial parts of Vitex negundo var. cannabifolia. The isolation was performed using bio-assay tracking experiments. The structures of compounds 1-5 were established by spectroscopic means. The antibacterial activities of the compounds were assessed against Escherichia coli, Bacillus subtilis, Micrococcus tetragenus and Pseudomonas fluorescens. Chrysosplenol-D (4) exhibited activities against all the four spoilage microorganisms.
Product Description
Isovitexin, primary reference standard
Sigma-Aldrich
Isovitexin, ≥98.0% (HPLC)