packaging
pack of 180 ea
manufacturer/tradename
TPP 87050
capacity
50 mL
size
General description
TPP® TubeSpin bioreactor tubes are ideal for large scale screening and optimizing of suspension cell system process development. The crucial parameters for the production upscaling can be analyzed in small amounts. This makes it cost-effective and increases the process efficiency.
Features and Benefits
- Available in conical or round bottom.
- Filter cap with partly 5 different openings (A, B, C, D, E) or 10 equal openings.
- Centrifuge apparatus which is fit standard.
- Spin: 15′500 x g or 3′500 x g.
- Provides matching adapter plates for shakers for the cultivation of suspension cells.
Legal Information
T3P is a registered trademark of Archimica GmbH
TPP is a registered trademark of TPP Techno Plastic Products AG
Science advances, 6(27) (2020-09-17)
Around 40% of humans and Tasmanian devils (Sarcophilus harrisii) develop cancer in their lifetime, compared to less than 10% for most species. In addition, devils are affected by two of the three known transmissible cancers in mammals. Immune checkpoint immunotherapy