1,000µL tips are recommended for this pipette. The manufacturer suggests product Z709956 which has a range of 10-1000µL.
Z757314
Sartorius mLINE® mechanical pipettor
single channel, volume 100-1000 μL
Synonym(s):
Multichannel pipettor
About This Item
Recommended Products
feature
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 725070
volume
100-1000 μL
General description
- Extremely light pipetting and tip ejection
- Unique safety features
- Stepless volume adjustment and patented volume locking system
- Thermal insulation of internal components to improve accuracy
- Full autoclavability without disassembling
- Easy cleaning, maintenance and calibration
- ID tag and color-coded caps
- Safe-Cone Filters and unique ejection
- Opti-load tip mounting mechanism for even tip sealing and light tip ejection in multichannel pipettors
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
-
Does this pipettor use only 1000 μL pipette tips?
1 answer-
Helpful?
-