Key Documents
Z741769
Sigma-Aldrich
Sigma Aldrich® Dual Run & Blot Accessory
Blotting Cassette with Pad, L × W 9 cm × 9 cm
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
manufacturer/tradename
Sigma-Aldrich
L × W
9 cm × 9 cm
General description
Sigma Aldrich gels run best on our Dual Run and Blot unit. This unique apparatus permits the running of both 10x10cm and 8x10cm gels. The unit is available in dual and quad form enabling up to 4 gels to be run at one time.
Both units can also be used to complete Western blots. The unique electrode arrangement provides a gradient electric field which ensures that both small and large proteins migrate from the gel at a comparable rate.
Application
Dual Run & Blot cassettes simply hinge open to accept blotting pads and membranes. Convenitently color coded to ensure correct insertion into the Dual Run & Blot Core Module
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany