Key Documents
Z719498
Sorenson low binding Ultra G microtips
maximum volume 10 μL, pack of 1000 ea (Bag), non-sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
10 μL
sterility:
non-sterile
feature:
barrier: no
graduations
packaging:
pack of 1000 ea (Bag)
manufacturer/tradename:
Sorenson 23580T
material
colorless
polypropylene
sterility
non-sterile
feature
barrier: no
graduations
packaging
pack of 1000 ea (Bag)
manufacturer/tradename
Sorenson 23580T
maximum volume
10 μL
volume
10 μL
Related Categories
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for ultra-micro pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)