Skip to Content
Merck
All Photos(1)

Key Documents

Z370827

Sigma-Aldrich

Cuvette mixer

Square footprint

Synonym(s):

cuvettes, micro cuvette, plastic cuvette

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

description

for full-size spectrophotometer cells and fluorimeter cells

technique(s)

UV/Vis spectroscopy: suitable
photometry: suitable

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Up and down plunging motion provides thorough mixing in spectrophotometer cells. Clear acrylic with straight handle; two different "footprints" for full-size and semi-micro cells with 10 mm light path.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service