TMO4112-0500
Nalgene® sterile single-use Erlenmeyer flask with solid closure
PETG, bottle volume 500 mL, case of 12 ea
Synonym(s):
Nalgene Erlenmeyer flask, single use plastic flask
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.13
material
PETG
sterility
sterile
packaging
case of 12 ea
manufacturer/tradename
Nalgene 4112-0500
bottle volume
500 mL
Related Categories
General description
Save preparation time and avoid contamination risk by using Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Other Notes
Browse all Erlenmeyer flasks
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)