Skip to Content
Merck
All Photos(4)

Key Documents

SAB5900201

Sigma-Aldrich

Alpaca Anti-GFP, recombinant single domain Antibody

Recombinant Alpaca(VHH) Nano antibody

Synonym(s):

GFP, Green fluorescent protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

alpaca

Quality Level

recombinant

expressed in E. coli

conjugate

unconjugated

antibody form

affinity purified VHH antibody

clone

GE10, monoclonal

Assay

>95% (SDS-PAGE)

form

lyophilized solid

mol wt

13.7 kDa
~15-18 kDa (SDS PAGE under reducing condition)

purified by

immobilized metal affinity chromatography (IMAC)

species reactivity

SARS coronavirus

concentration

1 mg/mL

technique(s)

ELISA: 1:2000-1:10000
immunofluorescence: 1:100-1:600
immunoprecipitation (IP): 0.5-4.0 μg
western blot: 1:1000-1:5000

color

colorless

pH

( in PBS, pH 7.4)

isotype

VHH

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Green fluorescent protein (GFP) is a protein composed of 238 amino acid residues(26.9kDa) derived from the Jellyfish Aequorea victoria, which emits green light (emission peak at 509nm) when excited by blue light (excitation peak at 395nm). GFP has become an invaluable tool in cell biology research, since its intrinsic fluorescence can be visualized in living cells. EGFP contains the double-amino-acid substitutions Phe-64 to Leu and Ser-65 to Thr (previously published as GFPmut1; PMID: 8707053). In contrast to wtGFP, EGFP has a single, strong, red-shifted excitation peak at 488 nm. Alpaca derived anti-GFP VHH single-domain antibody (molecular weight: 13.7 kDa), expressed in E. coli under conditions free from animal derived components.It has a 6His tag at its C-terminal.
Single-domain antibody (sdAb), also called nano-antibody, is the variable region isolated from the antigen-binding variable domain (VHH) of camelid heavy-chain-only antibody. It lacks light chains and a first constant domain(CH1). This configuration confers a number of unique advantages over conventional monoclonal antibodies. The small size of just 15 kDa (2~4 nm), single-domain antibody is the smallest antigen-binding fragment that can easily penetrate tissues, allowing them to bind epitopes that are inacessible to conventional antibodies. These single-domain antibodies offer superior chemical and thermal stability and increased solubility than conventional antibodies. All our antibodies are tested to ensure the highest level of specificity, sensitivity and reproducibility. Alpaca derived anti-GFP VHH single-domain antibody (molecular weight: 13.7 kDa), expressed in E. coli under conditions free from animal derived components.

Specificity

Green fluorescence protein (GFP) and other derivatives of GFP

Immunogen

Recombinant full length GFP protein

Application

ELISA, WB, IP, IF

Packaging

50 µg per vial / 100 µg per vial

Physical form

Affinity purified VHH single domain antibody lyophilized from PBS, pH7.4

Reconstitution

Reconstitute the lyophilized antibody with sterile deionized water or with 50% glycerol (ACS grade or better) to a stock solution of 1 mg/mL. Solubilize for 30 to 60 minutes at room temperature with occasional gentle mixing. Avoid vigorous shaking or vortexing.

Storage and Stability

Stable for 1 year at 4°C from date of receipt.

Other Notes

Storage at 4°C; Do not freeze.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service