Skip to Content
Merck
All Photos(1)

Key Documents

MTCP81001S

MTC Bio Reservoir

capacity 100 mL, sterile, pack of 100 ea (individually wrapped)

Synonym(s):

reagent dish, reagent holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121522
NACRES:
NB.12

sterility

sterile

packaging

pack of 100 ea (individually wrapped)

manufacturer/tradename

MTC Bio P8100-1S

capacity

100 mL

Looking for similar products? Visit Product Comparison Guide

General description

Reservoir, 100ml sterile , Individually wrapped, 100/cs

Features and Benefits

  • Graduated inner walls
  • Micro-trough U-shaped bottoms
  • Pouring spouts on all 4 corners

Legal Information

MTC is a trademark of MTC Bio

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service