Skip to Content
Merck
All Photos(2)

Key Documents

GE17-1082-01

Phenyl Sepharose High Performance

Cytiva 17-1082-01, pack of 75 mL

Synonym(s):

Phenyl sepharose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

phenyl

packaging

pack of 75 mL

manufacturer/tradename

Cytiva 17-1082-01

matrix

6% cross-linked agarose

particle size

24-44 μm

avg. part. size

34 μm

cleaning in place

2-14

working range

3-13

suitability

suitable for bioprocess medium (BioProcess medium.)
suitable for bioprocess medium

General description

Phenyl Sepharose High Performance is an aromatic hydrophobic interaction chromatography (HIC) medium, designed for intermediate and polishing step purification steps when high resolution has priority.

Features and Benefits

  • Aromatic HIC medium designed for intermediate purification and polishing steps
  • Selectivity similar to Phenyl Sepharose 6 Fast Flow (low sub) but higher resolution due to the smaller 34 μm bead size
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • The hydrophilic nature of the base matrix ensures low levels of non-specific binding

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Optimize salt conditions for protein binding and impurity passage in chromatography to achieve required selectivity.

Optimize salt conditions for protein binding and impurity passage in chromatography to achieve required selectivity.

Optimize salt conditions for protein binding and impurity passage in chromatography to achieve required selectivity.

Optimize salt conditions for protein binding and impurity passage in chromatography to achieve required selectivity.

See All

Protocols

Selectivity of HIC medium from Cytiva is based on properties such as hydrophobicity, porosity and particle size.

Here we discuss purification options when working with Sepharose® High Performance and we present some examples of media screening, capture and purification.

This page discusses various aspects of sample preparation for chromatographic purification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service