Skip to Content
Merck
All Photos(1)

Key Documents

G6670

Sigma-Aldrich

Anti-Growth Factor Independence-1 antibody, Mouse monoclonal

clone 2.5D.17, purified from hybridoma cell culture

Synonym(s):

Anti-GFI1, Anti-ZNF163

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

2.5D.17, monoclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa

species reactivity

rat, mouse, human

concentration

~2 mg/mL

technique(s)

immunohistochemistry: suitable
microarray: suitable
western blot: 1-2 μg/mL using total cell extract of A549 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GFI1(2672)
mouse ... Gfi1(14581)
rat ... Gfi1(24388)

General description

Monoclonal Anti-Growth Factor Independence-1 (GFI1) (mouse IgG1 isotype) is derived from the hybridoma 2.5D.17 produced by the fusion of mouse myeloma cells (SP2 cells) and splenocytes from BALB/c mice immunized with rat GFI1. Growth Factor Independence 1 (GFI1) is a zinc-finger transcriptional repressor, that is located on human chromosome 1p22. Gfi1 is highly expressed in thymus while Gfi1B is the predominant factor expressed in spleen, and both are expressed in bone marrow. It has a repression domain named SNAG at the N-terminal.† The protein also has a zinc finger DNA binding domain that is homologous to the domain in Gfi1B. Both bind to same consensus DNA sequence

Specificity

The antibody epitope resides within the GFI1 SNAG transcriptional-repression domain (amino acids 7-26).

Immunogen

rat growth factor independence-1 (GFI1).

Application

Monoclonal Anti-Growth Factor Independence-1 (GFI1) antibody may be used in:
  • immunoblotting
  • immunohistochemistry
  • EMSA (electrophoretic mobility shift analysis)

Biochem/physiol Actions

Growth factor independence-1 (Gfi1) plays a key role in hematopoiesis along with Gfi1B proteins. Cytokines that regulate innate immunity induce Gfi1 expression. Deficiency of this protein causes thymic and peripherial T lymphopenia and severe abnormalities in pre-T-cell development in mice. Mutations in this gene cause defects in T lymphocyte function and development in humans. Gfi1 autoregulates its expression in T lymphocytes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Analysis Note

The antibody may also be used in EMSA (electrophoretic mobility shift analysis).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loretta L Doan et al.
Journal of immunology (Baltimore, Md. : 1950), 170(5), 2356-2366 (2003-02-21)
T cell differentiation in the thymus is dependent upon signaling through the TCR and is characterized by the resulting changes in expression patterns of CD4 and CD8 surface coreceptor molecules. Although recent studies have characterized the effects of proximal TCR
Chinavenmeni S Velu et al.
Blood, 113(19), 4720-4728 (2009-03-13)
The zinc finger protein growth factor independent-1 (Gfi1) is a transcriptional repressor that is critically required for normal granulocytic differentiation. GFI1 loss-of-function mutations are found in some patients with severe congenital neutropenia (SCN). The SCN-associated GFI1-mutant proteins act as dominant
Targeted transcriptional repression of Gfi1 by GFI1 and GFI1B in lymphoid cells
Doan LL, et al.
Nucleic Acids Research, 32(8), 2508-2519 (2004)
Cloning of the human Gfi-1 gene and its mapping to chromosome region 1p22
Roberts T and Cowell JK
Oncogene, 14(8), 1003-1003 (1997)
Prognostic significance of high GFI1 expression in AML of normal karyotype and its association with a FLT3-ITD signature
Volpe G, et al.
Scientific reports, 7(1), 11148-11148 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service