EP5825739000
Eppendorf® Adapter
for 14 x Tube 5.0 mL, for rotor S-4-104, set of 2 pieces
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103911
NACRES:
NB.78
Recommended Products
feature
for 14 x Tube 5.0 mL, for rotor S-4-104
packaging
set of 2 pieces
manufacturer/tradename
Eppendorf® 5825739000
technique(s)
centrifugation: suitable
compatibility
for use with 750 mL round bucket
Related Categories
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service