Skip to Content
Merck
All Photos(1)

Key Documents

P3X63Ag8U.1

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

mouse blood

packaging

tube of 5 μg 94120802-DNA-5UG
pkg of vial of cells 94120802-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Myeloma

Cell Line Description

P3x63Ag8U.1 is a derivative of the P3x63Ag8 cell line (Sigma Catalogue number. 85011401) from the mouse myeloma MOPC21, which was derived from a female BALB/c mouse. This variant no longer synthesises gamma-1 chains but still produces kappa light chains which are not secreted.

Application

This cell line is useful for fusion to spleen cells to produce hybridomas or for hybridisation studies with other myelomas or lymphomas. Can also be used for immunoglobulin production.

Culture Medium

DMEM + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures at 5-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selection of the right fusion partner cell line is critical to obtain desired attributes of the hybridoma. Explore hybridoma fusion partners cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service