Skip to Content
Merck
All Photos(1)

Key Documents

C8739

Sigma-Aldrich

Cyclin-Dependent Kinase 5 human

recombinant, expressed in E. coli BL21, 50-100 units/μg protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli BL21

Quality Level

form

solution

specific activity

50-100 units/μg protein

packaging

vial of 300 units

shipped in

dry ice

storage temp.

−70°C

Application

Human cyclin-dependent kinase 5 has been used in a study to determine its 3′ untranslated region subunit 1 contains regulatory elements that affect transcript stability.

Biochem/physiol Actions

CDK5 is highly expressed in the nervous system where it phosphorylates a multitude of substrates. CDK5 plays an important role in many processes such as: apoptosis, phototransduction, dopamine and glutamate signalling, neurite outgrowth, neurone migration, induction of acetylcholine receptors, golgi membrane traffic and insulin exocytosis. It has also been implicated in the pathogenesis of amyotrophic lateral sclerosis . It is activated by p35 and p39, and their proteolytic cleavage products, p25 and p29.

Unit Definition

One unit will phosphorylate 1.0 picomole of the peptide substrate histone H19-18 per minute at 30 °C at pH 7.4.

Physical form

Supplied as a solution in PBS pH 7.4, 1 mM EDTA, 1mM DTT and 1 mg/ml BSA

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacological inhibitors of cyclin-dependent kinases
Knockaert M, et al.
Trends in Pharmacological Sciences, 23(9), 417-425 (200)
Kwok-On Lai et al.
The Journal of biological chemistry, 290(23), 14637-14646 (2015-04-24)
The maturation and maintenance of dendritic spines depends on neuronal activity and protein synthesis. One potential mechanism involves mammalian target of rapamycin, which promotes protein synthesis through phosphorylation of eIF4E-binding protein and p70 ribosomal S6 kinase 1 (S6K). Upon extracellular
Silvia Moncini et al.
BMC molecular biology, 8, 111-111 (2007-12-07)
CDK5R1 plays a central role in neuronal migration and differentiation during central nervous system development. CDK5R1 has been implicated in neurodegenerative disorders and proposed as a candidate gene for mental retardation. The remarkable size of CDK5R1 3'-untranslated region (3'-UTR) suggests
Shu Zhang et al.
PloS one, 10(7), e0131833-e0131833 (2015-07-07)
Cyclin-dependent kinase 5 (CDK5) is a cytoplasmic serine/ threonine kinase. Knockdown of CDK5 enhances paclitaxel sensitivity in human ovarian cancer cells. This study explores the mechanisms by which CDK5 regulates paclitaxel sensitivity in human ovarian cancers. Multiple ovarian cancer cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service