BAB602150000
H-B® Durac® Thermometer
temperature -20 - 150 °C (0-300 F), Maximum Registering, calibration
All Photos(1)
About This Item
Recommended Products
description
usage autoclave thermometer
feature
calibration
Maximum Registering
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art B60215-0000
temperature measuring range
-20 - 150 °C (0-300 F)
General description
Durable bi-metal thermometer has a maximum registering pointer (red) that is moved up by the temperature measuring pointer (black) to indicate the highest temperature achieved, and remains there until manually reset.
Features and Benefits
- Threaded connection (1/4” NPT) screws directly into in-line processes; can be used to mount thermometer on included stand
- Waterproof and pressurized; withstands autoclave cycles and has a removable protective silicone sleeve to prevent moisture from making contact with the dial
- Constructed from durable 304 stainless steel with a glass lens; withstands pressure, high temperatures and rust
- Probe length: 50mm (2")
Legal Information
Durac is a registered trademark of SP Scienceware, a division of SP
H-B is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)