SMB00945
Dihydroresveratrol
≥95% (HPLC)
Synonym(s):
Dihydroresveratrol, 3,4′,5-Trihydroxybibenzyl, 3,5,4′-Trihydroxybibenzyl
About This Item
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
mol wt
230.26 g/mol
storage condition
(Tightly closed. Dry.)
color
off-white to white
solubility
ethanol: 20 mg/mL
methanol: 20 mg/mL
storage temp.
2-8°C
SMILES string
Oc1cc(cc(c1)CCc2ccc(cc2)O)O
InChI
1S/C14H14O3/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(17)8-11/h3-9,15-17H,1-2H2
InChI key
HITJFUSPLYBJPE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service