BCR113
Potassium chloride fertilizer (elemental composition)
BCR®, certified reference material, elemental composition fertilizer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
product name
Potassium chloride fertilizer (elemental composition), BCR®, certified reference material
grade
certified reference material
Agency
BCR®
manufacturer/tradename
JRC
application(s)
agriculture
environmental
format
matrix material
Analysis Note
For more information please see:
BCR113
BCR113
Legal Information
BCR is a registered trademark of European Commission
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service