Skip to Content
Merck
All Photos(1)

Key Documents

94117

Supelco

Antimony Standard for AAS

TraceCERT®, 1 g/L Sb in nitric acid (nominal concentration)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1 g/L Sb in nitric acid (nominal concentration)

technique(s)

AAS: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Antimony standard may be used in the quantitative determination of antimony in geological samples using inductively coupled mass spectrometry and flow injection hydride generation atomic absorption spectrophotometry.[1]

Analysis Note

1 g/L Sb in 2% nitric acid (contains HF traces), prepared with high purity Sb metal, HNO3, HF and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates, please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulla B Hellström et al.
Virology journal, 6, 7-7 (2009-01-22)
Sci-B-Vac is a recombinant, hepatitis B vaccine derived from a mammalian cell line and containing hepatitis B surface antigen (HBsAg) as well as preS1 and preS2 antigens. Few studies have been performed on the antibody responses to preS1 in relation
Quantitative Determination of Antimony, Bismuth, and Tellurium in Geological Samples by Inductively Coupled Plasma Mass Spectrometry and Flow Injection Hydride Generation Atomic Absorption Technique
Saha.CM, et al.
Atomic Spectroscopy, 38, 7-11 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service