Skip to Content
Merck
All Photos(1)

Key Documents

399275

Sigma-Aldrich

Nω-Hydroxy-nor-L-arginine, Diacetate Salt

A potent, selective, competitive, and high affinity inhibitor of arginase.

Synonym(s):

Nω-Hydroxy-nor-L-arginine, Diacetate Salt, nor-NOHA, 2CH₃CO₂H, L-2-Amino-(4-(2ʹ-hydroxyguanidino)butyric Acid, 2CH₃CO₂H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N4O3 · 2C2H4O2
CAS Number:
Molecular Weight:
296.28
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (TLC)

form

solid

potency

2 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

off-white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N(O)\C(=N\CC[C@H](N)C(=O)O)\N.OC(=O)C.OC(=O)C

InChI key

PQFYTZJPYBMTCT-QTNFYWBSSA-N

General description

A potent, selective, competitive, and high affinity inhibitor of arginase. Shown to inhibit arginase from rat liver (IC50 = 2 µM) and mouse macrophages (IC50 = 50 µM). Shown to specifically interact with the manganese-cluster of the enzyme active site. Does not function as a substrate or as an inhibitor for NOS. Hence, nor-NOHA may be a useful tool to study the interplays between arginase and NOS.

Biochem/physiol Actions

Primary Target
arginase from rat liver
Secondary Target
mouse macrophages (IC₅₀ = 50 µM)

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Gobert, A.P., et al. 2000. Infect. Immun. 68, 4653.
Meurs, H., et al. 2000. Br. J. Pharmacol. 130, 1793.
Moali, C., et al. 2000. Biochemistry 39, 8208.
Tenu, J.P. et al. 1999. Nitric Oxide 3, 427.
Moali, C., et al. 1998. Biochemistry 37, 10453.
Custot, J., et al. 1997. J. Am. Chem. Soc. 119, 4086.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Zhu et al.
Investigative ophthalmology & visual science, 62(7), 3-3 (2021-06-02)
To investigate the effects of ex vivo-induced bone marrow myeloid-derived suppressor cells (BM-MDSCs) on allogeneic immune responses in corneal transplantation. Bone marrow cells from C57BL/6J (B6) mice were cultured with IL-6 and GM-CSF for four days. The ex vivo induction
Rashi Sehgal et al.
Frontiers in immunology, 13, 828949-828949 (2022-06-21)
Decompensated cirrhosis patients are more prone to bacterial infections. Myeloid-derived suppressor cells (MDSCs) expand in sepsis patients and disrupt immune cell functions. Granulocyte-macrophage colony-stimulating factor (GM-CSF) therapy helps in restoring immune cell functions and resolving infections. Its role in MDSC
Alessandra Sacchi et al.
Cell death & disease, 11(10), 921-921 (2020-10-29)
The immunological mechanisms underlying the clinical presentation of SARS-CoV-2 infection and those influencing the disease outcome remain to be defined. Myeloid-derived suppressor cells (MDSC) have been described to be highly increased during COVID-19, however, their role remains elusive. We performed
Máté Kiss et al.
Cancer immunology research, 9(3), 309-323 (2020-12-29)
IL1β is a central mediator of inflammation. Secretion of IL1β typically requires proteolytic maturation by the inflammasome and formation of membrane pores by gasdermin D (GSDMD). Emerging evidence suggests an important role for IL1β in promoting cancer progression in patients

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service