Skip to Content
Merck
All Photos(1)

Key Documents

385920

Sigma-Aldrich

HSP90 Inhibitor, CCT018159

The HSP90 Inhibitor, CCT018159, also referenced under CAS 171009-07-7, controls the biological activity of HSP90.

Synonym(s):

HSP90 Inhibitor, CCT018159, 4-(4-(2,3-Dihydro-1,4-benzodioxin-6-yl)-5-methyl-1H-pyrazol-3-yl)-6-ethylresorcinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20N2O4
CAS Number:
Molecular Weight:
352.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 1.7 mg/mL
DMSO: 35 mg/mL
ethanol: 35 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[nH]1nc(c(c1C)c3cc4c(cc3)OCCO4)c2c(cc(c(c2)CC)O)O

InChI

1S/C20H20N2O4/c1-3-12-8-14(16(24)10-15(12)23)20-19(11(2)21-22-20)13-4-5-17-18(9-13)26-7-6-25-17/h4-5,8-10,23-24H,3,6-7H2,1-2H3,(H,21,22)

InChI key

OWPMENVYXDJDOW-UHFFFAOYSA-N

General description

A cell-permeable pyrazoloresorcinol compound that inhibits HSP90 ATPase activity (IC50 = 3.2 µM and 6.6 µM against human HSP90β and yeast HSP90, respectively) by targeting the enzyme′s N-terminal ATP binding site. It exhibits no detectable effects against human Hsp70 or topoisomerase II even at concentrations as high as 100 µM and inhibits a panel of 20 commonly studied kinases only at much higher concentrations. Unlike 17-AAG (Cat. No. 100068), the antitumor activity of CCT018159 is not affected by the expression of NQ01/DT-diaphorase or P-glycoprotein.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solution are stable for up to 3 months at -20°C.

Other Notes

Hardcastle, A., et al. 2007. Mol. Cancer Ther.6, 1112.
Sharp, S.Y., et al. 2007. Cancer Res.67, 2206.
Smith, N.F., et al. 2006. Mol. Cancer Ther.5, 1628.
Dymock, B.W., et al. 2005. J. Med. Chem.48, 4212.
Cheung, K.M., et al. 2005. Bioorg. Med. Chem. Lett.15, 3338.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service