Skip to Content
Merck
All Photos(1)

Key Documents

310500

Sigma-Aldrich

D-erythro-Sphingosine, N,N-Dimethyl-

A cell-permeable and reversible inhibitor of protein kinase C (PKC; IC₅₀ = 12 µM) and stimulates Src kinase activity.

Synonym(s):

D-erythro-Sphingosine, N,N-Dimethyl-, DMS, SK Inhibitor III, Sphingosine Kinase Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H41NO2
CAS Number:
Molecular Weight:
327.55
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

oil (Clear)
waxy solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

DMSO: 5 mg/mL
ethanol: soluble
methanol: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

N([C@H]([C@H](O)\C=C\CCCCCCCCCCCCC)CO)(C)C

InChI

1S/C20H41NO2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(23)19(18-22)21(2)3/h16-17,19-20,22-23H,4-15,18H2,1-3H3/b17-16+/t19-,20+/m0/s1

InChI key

YRXOQXUDKDCXME-YIVRLKKSSA-N

General description

A cell-permeable and reversible inhibitor of protein kinase C (PKC; IC50 = 12 µM) and stimulates Src kinase activity. Useful for inhibiting cell surface expression of selectins that promote adhesion of leukocytes or tumor cells to platelets and endothelial cells. Induces apoptosis in human leukemia HL-60 cells. An inhibitor of sphingosine kinase.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC
Product does not compete with ATP.
Reversible: yes
Target IC50: 12 µM against PKC

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 weeks at -20°C.

Other Notes

Cuvillier, O., et al. 1996. Nature381, 800.
Ohta, H., et al. 1995. Cancer Res.55, 691.
Kimura, S., et al. 1992. Biochem. Pharmacol.44, 1585.
Hanada, K., et al. 1991. Biochemistry30, 11682.
Igarashi, Y., et al. 1990. J. Biol. Chem.265, 5385.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grégory Lavieu et al.
Methods in molecular biology (Clifton, N.J.), 445, 159-173 (2008-04-22)
Sphingolipids are constituents of biological membranes. Ceramide and sphingosine 1-phosphate (S1P) also act as second messengers and are part of a rheostat system, in which ceramide promotes cell death and growth arrest, and S1P induces proliferation and maintains cell survival.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service