Skip to Content
Merck
All Photos(1)

Documents

1.16720

Supelco

Glucose Test Strips, reflectometric

1-100 mg/L (glucose), REFLECTOQUANT®

Synonym(s):

Glucose chemical test strips, Glucose test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Glucose Test, reflectometric, 1-100 mg/L (glucose), for use with REFLECTOQUANT®

description

quantitative

Quality Level

analyte chemical class(es)

sugars

measuring range

1-100 mg/L (glucose)

compatibility

RQflex® 10 Plus
RQflex® 10
RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips 1 - 100 mg/l Reflectoquant®
The Reflectoquant Glucose test is suitable for determining glucose in beverages, food and preserves. Furthermore you can use it in fermentation solutions. Under the catalytic effect of glucose oxidase, glucose is converted into gluconic acid lactone. In the presence of peroxidase the hydrogen peroxide formed in the process reacts with an organic redox indicator to form a blue-green dye that is determined reflectometrically. For pre-testing a sample in an economic and quick way to estimate the glucose concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strip for glucose.

Application


  • Bifunctional oxidase-peroxidase mimicking Fe-Ce MOF on paper-based analytical devices to intensify luminol chemiluminescence: This research introduces an innovative glucose assay technique using MOF-enhanced paper devices, providing a cost-effective and accessible method for glucose monitoring in resource-limited settings (Hassanzadeh et al., 2024).

  • Zinc and Silicon Nano-Fertilizers Influence Ionomic and Metabolite Profiles in Maize to Overcome Salt Stress: In this agricultural research, glucose tests are used to monitor the health and metabolic responses of maize to nano-fertilizer treatment, demonstrating the versatility of glucose assays in environmental and plant science applications (Shoukat et al., 2024).

Linkage

Replaces: 16720; 16720-1

Analysis Note

function control: conforms
Assay
- standard 1:
9 - 11 mg/l
- standard 2: 36 - 44 mg/l
- standard 3: 72 - 88 mg/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyunjin Kim et al.
Bioresource technology, 301, 122794-122794 (2020-01-26)
Caproic acid (CA) was produced by Megasphaera elsdenii T81 with Jerusalem artichoke tubers (JA) as a feedstock. More CA was produced under the medium with the acid hydrolysate of JA than the comparative medium with a carbon composition similar to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service