Key Documents
Z693731
LabJaws jumbo clamp holder
stainless steel
Synonym(s):
clamp holders, frame connectors
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
Pricing and availability is not currently available.
material
stainless steel
manufacturer/tradename
Troemner 916357
jaw capacity
0-18 mm
Related Categories
General description
Corrosion resistant. Has precise fit and powerful holding ability for heavy-duty applications. Ideal for holding clamps to lab frames or ring stands.
Legal Information
LabJaws is a trademark of Henry Troemner LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)