Key Documents
Z261939
Optifix® PTFE thread adapter
neck thread size GL 38
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
Pricing and availability is not currently available.
neck thread size
GL 38
manufacturer/tradename
Fortuna Graf
availability
not available in EU
General description
Made of PTFE. Adapts Optifix dispensers to other bottle thread sizes.
Legal Information
Optifix is a registered trademark of Poulten & Graf Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)