SYND681922
Synthware™ distillation distribution adapter with vacuum hose connection
joint: ST/NS 19/22
Synonym(s):
Synthware D681922, radial style distillation receiver
All Photos(1)
About This Item
UNSPSC Code:
41104812
NACRES:
NB.51
Recommended Products
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware D681922
Related Categories
General description
Cow-type design having three receivers spaced 40° apart. 105° angle between top outer joint and lower inner joints.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)