MT51302080
Mettler-Toledo Rainbow II buffer
pH 4.01/7.00/10.01
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
51191600
NACRES:
NB.79
Recommended Products
sterility
non-sterile
form
liquid
manufacturer/tradename
Mettler-Toledo 51302080
packaging
pkg of (3 packs 10 x 20 mL sachets)
pH
4.01/7.00/10.01
General description
Mettler-Toledo Rainbow II buffers promise a fresh and sterilized solution for every calibration. It is available as single-use sachets for effective calibration under controlled conditions. The box design allows one to pick one sachet at a time, single-handedly and with ease. It comprises sachets for 10 calibrations.
Features and Benefits
- Minimizes contamination risks
- Round and large opening of the sachets allows easier sensor insertion
- Calibration is made a quick and simple task
- Effortless sachet dispensing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service