Skip to Content
Merck
All Photos(2)

Key Documents

GF32867893

Lead

foil, 8mm disks, thickness 0.1mm, as rolled, 99.95%

Synonym(s):

Lead, PB000280

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 328-678-93

resistivity

20.65 μΩ-cm

diam. × thickness

8 mm × 0.1 mm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lead foil in the intraoral periapical radiograph as an inclusion agent: a simple method in denture identification.
Thorakkal Shamim
Journal of the College of Physicians and Surgeons--Pakistan : JCPSP, 22(2), 130-131 (2012-02-09)
Motoaki Kato et al.
Nihon Hoshasen Gijutsu Gakkai zasshi, 61(8), 1158-1169 (2005-09-01)
In projection radiography, stationary grids are indispensable accessories to the improvement of diagnostic imaging. On the other hand, they are becoming one of the issues facing digital image processing. The lead foil that composes the grid can produce moiré on
M Bourgeois et al.
Health physics, 62(6), 546-552 (1992-06-01)
A significant amount of radiation escapes the rear of an image receptor in conventional dental radiographic procedures despite the placement of a lead-containing backing at the rear of the film pack. The purpose of this investigation was to place additional
Lizhi Zhang et al.
Small (Weinheim an der Bergstrasse, Germany), 1(3), 349-354 (2006-12-29)
In this study we demonstrate that one-dimensional (1D) nanostructured lead telluride (PbTe) can be synthesized in a hydrothermal reaction between lead foil and tellurium powder. The resulting materials were characterized by X-ray diffraction, scanning electron microscopy, and transmission electron microscopy.
D W Rogers
Medical physics, 26(4), 533-537 (1999-05-05)
Data are presented to allow the photon beam quality specifier being used in the new AAPM TG-51 protocol, %dd(10)x, to be extracted from depth-dose data measured with a 1 mm lead foil either 50 cm or 30 cm from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service