BAF369110025
SP Bel-Art® Sterileware™ Flat Bottom Sampling Scoop
white polystyrene, nominal capacity 25 mL (0.8 oz), sterile, pack of 100 ea (individually wrapped)
Synonym(s):
collection scoop
Sign Into View Organizational & Contract Pricing
All Photos(6)
About This Item
Recommended Products
material
white polystyrene
sterility
sterile
packaging
pack of 100 ea (individually wrapped)
manufacturer/tradename
Bel-Art F36911-0025
bowl H × L × W
0.75 in. (20 mm) × 2.625 in. (67 mm) × 1.5 in. (38 mm)
handle L
2.75 in. (7 cm)
nominal capacity
25 mL (0.8 oz)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
These sterile scoops are used by pharmaceutical, food, cosmetic and chemical companies for general production or laboratory work.
Features and Benefits
These sterile scoops are used by pharmaceutical, food, cosmetic and chemical companies for general production or laboratory work.
Note: Sterilized products are not returnable.
These sampling scoops are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes.
- Handle bump acts as thumb rest to help keep the hand away from the scoop; reduces contamination risks
- Flat bottom allows for use as a weigh boat
- Approximate volumes can be measured by removing excess powder
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Ready for one time use and disposal
- Lot stamped for accurate tracing
Note: Sterilized products are not returnable.
These sampling scoops are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service