738611
5-(Dicyclohexylphosphino)-1′,3′,5′-triphenyl-1′H-[1,4′]bipyrazole
97%
Synonym(s):
Cy-BippyPhos
About This Item
Quality Level
Assay
97%
form
solid
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
mp
177-180 °C (lit.)
functional group
phosphine
storage temp.
2-8°C
SMILES string
C1CCC(CC1)P(C2CCCCC2)c3ccnn3-c4c(nn(-c5ccccc5)c4-c6ccccc6)-c7ccccc7
InChI
1S/C36H39N4P/c1-6-16-28(17-7-1)34-36(35(29-18-8-2-9-19-29)39(38-34)30-20-10-3-11-21-30)40-33(26-27-37-40)41(31-22-12-4-13-23-31)32-24-14-5-15-25-32/h1-3,6-11,16-21,26-27,31-32H,4-5,12-15,22-25H2
InChI key
ZMENEJKATASVIH-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service