About This Item
Recommended Products
grade
technical grade
mp
58-60 °C (lit.)
storage temp.
2-8°C
SMILES string
COc1ccc(C(Cl)=O)c(OC)c1
InChI
1S/C9H9ClO3/c1-12-6-3-4-7(9(10)11)8(5-6)13-2/h3-5H,1-2H3
InChI key
KVIUXRJCBBXEGJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service