About This Item
Recommended Products
vapor pressure
5 mmHg ( 94 °C)
Assay
97%
form
liquid
reaction suitability
core: aluminum
bp
108-109 °C/10 mmHg (lit.)
mp
2.5-4.5 °C (lit.)
density
0.85 g/mL at 25 °C (lit.)
SMILES string
CCO[Al](CC)CC
InChI
1S/C2H5O.2C2H5.Al/c1-2-3;2*1-2;/h2H2,1H3;2*1H2,2H3;/q-1;;;+1
InChI key
GCPCLEKQVMKXJM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Compatible with the following:
- Aldrich® Sure/Pac™ station for liquefied gases Z566446
- PTFE Sealing tape Z104388 or Z221880
- Straight septum-inlet adapter Z118141 with septa Z565687 or Z565695
Legal Information
also commonly purchased with this product
recommended
septum inlet adapter
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 1
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
nwg
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service