Merck
CNS stem cells express a new class of intermediate filament protein.
Cell (1990-02-23)
U Lendahl, L B Zimmerman, R D McKay
ABSTRACT
Multipotential CNS stem cells receive and implement instructions governing differentiation to diverse neuronal and glial fates. Exploration of the mechanisms generating the many cell types of the brain depends crucially on markers identifying the stem cell state. We describe a gene whose expression distinguishes the stem cells from the more differentiated cells in the neural tube. This gene was named nestin because it is specifically expressed in neuroepithelial stem cells. The predicted amino acid sequence of the nestin gene product shows that nestin defines a distinct sixth class of intermediate filament protein. These observations extend a model in which transitions in intermediate filament gene expression reflect major steps in the pathway of neural differentiation.
Sigma-Aldrich
Human Neural Stem Cell Characterization Kit, The Human Neural Stem Cell Characterization Kit contains three molecular markers, Nestin, Sox 2 & Musashi that are frequently used to identify neural stem/progenitor cells.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Neural Stem Cell Marker Characterization Kit, The Neural Stem Cell Characterization Kit contains two molecular markers, Nestin & Sox 2 that are frequently used to identify neural stem/progenitor cells.