Merck
Development of Next Generation Stevia Sweetener: Rebaudioside M.
Foods (Basel, Switzerland) (2014-02-27)
Indra Prakash, Avetik Markosyan, Cynthia Bunders
ABSTRACT
This work aims to review and showcase the unique properties of rebaudioside M as a natural non-caloric potential sweetener in food and beverage products. To determine the potential of rebaudioside M, isolated from Stevia rebaudiana Bertoni, as a high potency sweetener, we examined it with the Beidler Model. This model estimated that rebaudioside M is 200-350 times more potent than sucrose. Numerous sensory evaluations of rebaudioside M's taste attributes illustrated that this steviol glycoside possesses a clean, sweet taste with a slightly bitter or licorice aftertaste. The major reaction pathways in aqueous solutions (pH 2-8) for rebaudioside M are similar to rebaudioside A. Herein we demonstrate that rebaudioside M could be of great interest to the global food industry because it is well-suited for blending and is functional in a wide variety of food and beverage products.
Sigma-Aldrich
Rebaudioside M, ≥95.0% (HPLC)